Assay Details
Target Gene Details
Entrez Gene ID: | 320571 |
Gene Name: | ATPase, class I, type 8B, member 5 |
Gene Aliases: |
4930417M19Rik, Feta |
Location: |
Chr.4:43266714-43373851 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp8b5 | NM_177195.3 | 24 | 2715 | NP_796169.2 |
| AK029992.1 | 25 | 2946 | ||
| EF377342.1 | 24 | 2704 |
More Information
Set Membership: |
|

Genomic Map