Assay Details
Target Gene Details
Entrez Gene ID: | 230883 |
Gene Name: | arylacetamide deacetylase-like 3 |
Gene Aliases: |
Gm435 |
Location: |
Chr.4:144453771-144463756 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aadacl3 | NM_001085503.2 | NP_001078972.1 |
More Information
Set Membership: |
|

Genomic Map