Assay Details
Target Gene Details
Entrez Gene ID: | 70093 |
Gene Name: | ubiquitin-conjugating enzyme E2Q (putative) 1 |
Gene Aliases: |
1110002C01Rik, 2310012M18Rik, AI854014, Gtap, NICE-5, PRO3094, Ube2q |
Location: |
Chr.3:89773609-89783997 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ube2q1 | NM_027315.4 | 3 | 587 | NP_081591.3 |
| AK009324.1 | 2 | 169 | BAB26217.2 | |
| AK146707.1 | 3 | 400 | BAE27373.1 | |
| AK177704.1 | 2 | 139 | ||
| AK214106.1 | 2 | 108 | ||
| AY112699.1 | 3 | 580 | AAM60815.1 | |
| AY945937.1 | 3 | 484 | AAY23286.1 | |
| BC060966.1 | AAH60966.1 | |||
| BC082275.1 | 3 | 410 | AAH82275.1 |
More Information
Set Membership: |
|

Genomic Map