Assay Details
Target Gene Details
Entrez Gene ID: | 21427 |
Gene Name: | vacuolar protein sorting 72 (yeast) |
Gene Aliases: |
Tcfl1, YL-1 |
Location: |
Chr.3:95111042-95123051 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps72 | NM_009336.2 | NP_033362.2 | ||
| AK140284.1 | BAE24317.1 | |||
| AK155191.1 | BAE33106.1 | |||
| AK179387.1 | ||||
| BC004834.1 | AAH04834.1 | |||
| BC043029.1 | AAH43029.2 | |||
| BY294768.1 | ||||
| D43643.1 | BAA07758.1 |
More Information
Set Membership: |
|

Genomic Map