Assay Details
Target Gene Details
Entrez Gene ID: | 69073 |
Gene Name: | keratinocyte differentiation factor 1 |
Gene Aliases: |
1810019J16Rik |
Location: |
Chr.4:133518963-133530790 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdf1 | NM_001083916.1 | NP_001077385.1 | ||
| NM_133707.2 | NP_598468.2 | |||
| AK007551.1 | BAC25177.1 | |||
| AK158383.1 | ||||
| BC006890.1 | AAH06890.1 |

Genomic Map