Assay Details
Target Gene Details
Entrez Gene ID: | 433702 |
Gene Name: | nuclear cap binding protein subunit 1 |
Gene Aliases: |
AU014645, AW538051, CBP80 |
Location: |
Chr.4:46138511-46172403 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncbp1 | NM_001033201.3 | 5 | 771 | NP_001028373.2 |
| AK133991.1 | 5 | 728 | ||
| AK134334.1 | 5 | 762 | BAE22102.1 | |
| AK166458.1 | 5 | 553 | BAE38789.1 | |
| AK169557.1 | 5 | 772 | BAE41227.1 | |
| BC055777.1 | 2 | 197 | AAH55777.1 | |
| BC138898.1 | 5 | 642 |
More Information
Set Membership: |
|

Genomic Map