Assay Details
Target Gene Details
Entrez Gene ID: | 70533 |
Gene Name: | basic transcription factor 3-like 4 |
Gene Aliases: |
4632412E09Rik, 5730434I03Rik, AI316863, AU022207, AW552446 |
Location: |
Chr.4:108814295-108834297 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btf3l4 | NM_027453.2 | NP_081729.1 | ||
| AK004239.1 | BAB23233.1 | |||
| AK011367.1 | BAB27573.1 | |||
| AK013120.1 | BAB28660.1 | |||
| AK017614.1 | ||||
| AK076283.1 | BAC36287.1 | |||
| BC024612.1 | AAH24612.2 | |||
| BC058282.1 | AAH58282.1 | |||
| BC099407.1 | AAH99407.1 | |||
| BC138215.1 | ||||
| BC138216.1 |
More Information
Set Membership: |
|

Genomic Map