Assay Details
Target Gene Details
Entrez Gene ID: | 67490 |
Gene Name: | UFM1 specific ligase 1 |
Gene Aliases: |
1810074P20Rik, AI429228, Kiaa0776, Maxer, Rcad, mKIAA0776 |
Location: |
Chr.4:25247820-25281926 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ufl1 | NM_026194.4 | 18 | 2145 | NP_080470.2 |
| AK007993.2 | 18 | 2090 | BAB25395.1 | |
| AK013382.1 | 11 | 1407 | BAC25404.1 | |
| AK032663.1 | 18 | 2081 | BAC27976.1 | |
| AK082785.1 | 8 | 852 | BAC38618.1 | |
| AK129211.1 | 17 | 2058 | BAC98021.1 | |
| AK132697.1 | 18 | 2109 | BAE21308.1 | |
| BC132195.1 | 18 | 2097 | ||
| BC138153.1 | 18 | 2097 |
More Information
Set Membership: |
|

Genomic Map