Assay Details
Target Gene Details
Entrez Gene ID: | 30924 |
Gene Name: | angiopoietin-like 3 |
Gene Aliases: |
hypl |
Location: |
Chr.4:99030955-99038188 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Angptl3 | NM_013913.3 | NP_038941.1 | ||
| AF162224.1 | AAD45920.1 | |||
| AK149450.1 | BAE28886.1 | |||
| AK149555.1 | BAE28956.1 | |||
| AK163595.1 | ||||
| BC019491.1 | AAH19491.1 |
Target Gene Details
Entrez Gene ID: | 67299 |
Gene Name: | dedicator of cytokinesis 7 |
Gene Aliases: |
3110056M06Rik, Gm430, m |
Location: |
Chr.4:98936659-99122605 on Build GRCm38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dock7 | NM_001290636.1 | NP_001277565.1 | ||
| NM_026082.5 | NP_080358.3 | |||
| CA752561.1 | ||||
| DQ109674.1 | AAZ17650.1 |
More Information
Set Membership: |
|

Genomic Map