Assay Details
Target Gene Details
Entrez Gene ID: | 12259 |
Gene Name: | complement component 1, q subcomponent, alpha polypeptide |
Gene Aliases: |
AI255395, C1q |
Location: |
Chr.4:136895916-136898844 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1qa | NM_007572.2 | NP_031598.2 | ||
| AK002655.1 | BAB22262.1 | |||
| AK136737.1 | BAE23115.1 | |||
| AK150771.1 | BAE29837.1 | |||
| AK159210.1 | BAE34901.1 | |||
| AK159343.1 | BAE35005.1 | |||
| BC002086.1 | AAH02086.1 | |||
| BY318890.1 |

Genomic Map