Assay Details
Target Gene Details
Entrez Gene ID: | 12259 |
Gene Name: | complement component 1, q subcomponent, alpha polypeptide |
Gene Aliases: |
AI255395, C1q |
Location: |
Chr.4:136895916-136898844 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1qa | NM_007572.2 | 2 | 229 | NP_031598.2 |
| AK002655.1 | 2 | 155 | BAB22262.1 | |
| AK136737.1 | 2 | 155 | BAE23115.1 | |
| AK150771.1 | 2 | 155 | BAE29837.1 | |
| AK159210.1 | 2 | 155 | BAE34901.1 | |
| AK159343.1 | 2 | 150 | BAE35005.1 | |
| BC002086.1 | 2 | 188 | AAH02086.1 | |
| BY318890.1 | 2 | 232 | ||
| X58861.1 | 1 | 152 | CAA41664.1 |

Genomic Map