Assay Details
Target Gene Details
Entrez Gene ID: | 12260 |
Gene Name: | complement component 1, q subcomponent, beta polypeptide |
Gene Aliases: |
- |
Location: |
Chr.4:136880145-136886177 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1qb | NM_009777.2 | NP_033907.1 | ||
| AK140712.1 | BAE24452.1 | |||
| AK152764.1 | BAE31477.1 | |||
| AK153124.1 | BAE31739.1 | |||
| BC067001.1 | AAH67001.1 | |||
| M22531.1 | AAA37335.1 | |||
| M36293.1 | AAA37283.1 | |||
| X16874.1 | CAA34757.1 |

Genomic Map