Assay Details
Target Gene Details
Entrez Gene ID: | 107995 |
Gene Name: | cell division cycle 20 |
Gene Aliases: |
2310042N09Rik, C87100, p55CDC |
Location: |
Chr.4:118432901-118437343 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc20 | NM_023223.2 | 10 | 1712 | NP_075712.2 |
| AB045313.1 | 10 | 1616 | BAA97451.1 | |
| AF312208.1 | 10 | 1609 | AAL25714.1 | |
| AK011131.1 | 8 | 1099 | ||
| AK011723.1 | 10 | 1711 | ||
| AK075998.1 | 10 | 1660 | BAC36109.1 | |
| AK076030.1 | 10 | 1664 | BAC36132.1 | |
| AK083459.1 | 10 | 1541 | BAC38922.1 | |
| AK160204.1 | 10 | 1724 | ||
| AK168650.1 | 10 | 1664 | BAE40507.1 | |
| AV499208.1 | 1 | 171 | ||
| BC003215.1 | 10 | 1649 | AAH03215.1 |
Target Gene Details
Entrez Gene ID: | 54325 |
Gene Name: | elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 1 |
Gene Aliases: |
AA407424, BB151133, Ssc1 |
Location: |
Chr.4:118428081-118432953 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl1 | NM_001039175.2 | NP_001034264.1 | ||
| NM_001039176.2 | NP_001034265.1 | |||
| NM_019422.3 | NP_062295.1 | |||
| BM445297.1 | ||||
| BY689799.1 |
More Information
Set Membership: |
|

Genomic Map