Assay Details
Target Gene Details
Entrez Gene ID: | 107995 |
Gene Name: | cell division cycle 20 |
Gene Aliases: |
2310042N09Rik, C87100, p55CDC |
Location: |
Chr.4:118432901-118437343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc20 | NM_023223.2 | NP_075712.2 | ||
| AB045313.1 | BAA97451.1 | |||
| AF312208.1 | AAL25714.1 | |||
| AK011131.1 | ||||
| AK011723.1 | ||||
| AK029424.1 | ||||
| AK075998.1 | BAC36109.1 | |||
| AK076030.1 | BAC36132.1 | |||
| AK083459.1 | BAC38922.1 | |||
| AK160204.1 | ||||
| AK168650.1 | BAE40507.1 | |||
| AK188208.1 | ||||
| AK200715.1 | ||||
| AK205599.1 | ||||
| AK205741.1 | ||||
| AK206746.1 | ||||
| AK206928.1 | ||||
| AK207342.1 | ||||
| AK207468.1 | ||||
| AK208541.1 | ||||
| AK208620.1 | ||||
| AK208689.1 | ||||
| AK209126.1 | ||||
| AK209399.1 | ||||
| AK210837.1 | ||||
| AK211703.1 | ||||
| AK213222.1 | ||||
| AK215738.1 | ||||
| AK215892.1 | ||||
| AK216377.1 | ||||
| AK217035.1 | ||||
| AK219314.1 | ||||
| AK219440.1 | ||||
| AK219805.1 | ||||
| AK219889.1 | ||||
| BC003215.1 | AAH03215.1 |
More Information
Set Membership: |
|

Genomic Map