Assay Details
Target Gene Details
Entrez Gene ID: | 22239 |
Gene Name: | UDP galactosyltransferase 8A |
Gene Aliases: |
AI850488, AW455908, Cgt, Ugt8, mCerGT |
Location: |
Chr.3:125865322-125938550 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt8a | NM_011674.4 | NP_035804.2 | ||
| AK128994.1 | ||||
| AK137364.1 | BAE23325.1 | |||
| AK144005.1 | ||||
| AK163650.1 | ||||
| X92122.1 | CAA63090.1 |

Genomic Map