Assay Details
Target Gene Details
Entrez Gene ID: | 67490 |
Gene Name: | UFM1 specific ligase 1 |
Gene Aliases: |
1810074P20Rik, AI429228, Kiaa0776, Maxer, Rcad, mKIAA0776 |
Location: |
Chr.4:25247820-25281926 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ufl1 | NM_026194.4 | NP_080470.2 | ||
| AK007993.2 | BAB25395.1 | |||
| AK032663.1 | BAC27976.1 | |||
| AK076206.1 | ||||
| AK129211.1 | BAC98021.1 | |||
| AK132697.1 | BAE21308.1 | |||
| AK207708.1 | ||||
| AK208712.1 | ||||
| AK218998.1 | ||||
| BC132195.1 | ||||
| BC138153.1 |
More Information
Set Membership: |
|

Genomic Map