Assay Details
Target Gene Details
Entrez Gene ID: | 550619 |
Gene Name: | AT rich interactive domain 3C (BRIGHT-like) |
Gene Aliases: |
OTTMUSG00000006683 |
Location: |
Chr.4:41723831-41731867 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arid3c | NM_001017362.2 | 4 | 606 | NP_001017362.1 |
| NM_001252622.1 | 4 | 606 | NP_001239551.1 | |
| BC141237.1 | 3 | 575 | ||
| BC145563.1 | 3 | 575 |
More Information
Set Membership: |
|

Genomic Map