Assay Details
Target Gene Details
Entrez Gene ID: | 66625 |
Gene Name: | PNN interacting serine/arginine-rich |
Gene Aliases: |
5730406M06Rik, AA673174, Pinsr, Sfrs18, Srsf18 |
Location: |
Chr.4:21847583-21876475 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnisr | NM_025669.1 | NP_079945.1 | ||
| AK012036.1 | ||||
| AK017507.1 | BAB30779.3 | |||
| AK020035.1 | ||||
| AK031964.1 | ||||
| AK081407.1 | ||||
| AK083021.1 | ||||
| AK160288.1 | BAE35727.1 | |||
| AK162695.1 | ||||
| BC141031.1 |
More Information
Set Membership: |
|

Genomic Map