Assay Details
Target Gene Details
Entrez Gene ID: | 272027 |
Gene Name: | thiosulfate sulfurtransferase (rhodanese)-like domain containing 2 |
Gene Aliases: |
3010020C06 |
Location: |
Chr.4:46114746-46138581 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tstd2 | NM_173033.3 | NP_766621.3 | ||
| AK028258.1 | BAC25846.1 | |||
| AK155456.1 | BAE33273.1 | |||
| BC057893.1 | AAH57893.1 | |||
| CD549731.1 |
More Information
Set Membership: |
|

Genomic Map