Assay Details
Target Gene Details
Entrez Gene ID: | 73721 |
Gene Name: | RIKEN cDNA 1110017D15 gene |
Gene Aliases: |
Cbe1, Smrp1 |
Location: |
Chr.4:41505005-41517333 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110017D15Rik | NM_001048005.1 | NP_001041470.1 | ||
| NM_001253777.1 | NP_001240706.1 | |||
| NM_001253778.1 | NP_001240707.1 | |||
| NM_001253780.1 | NP_001240709.1 | |||
| NM_001253788.1 | NP_001240717.1 | |||
| NM_028624.1 | NP_082900.1 | |||
| NR_045549.2 | ||||
| NR_045551.2 | ||||
| AB231294.1 | BAE78528.1 | |||
| AK003742.1 | BAB22974.1 | |||
| BU700151.1 | ||||
| CA463601.1 | ||||
| DQ873295.1 | ||||
| DQ873296.1 |

Genomic Map