Assay Details
Target Gene Details
Entrez Gene ID: | 22065 |
Gene Name: | transient receptor potential cation channel, subfamily C, member 3 |
Gene Aliases: |
Mwk, Trcp3, Trp3, Trrp3 |
Location: |
Chr.3:36620482-36690229 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpc3 | NM_019510.2 | NP_062383.2 | ||
| AF190645.1 | AAF01468.1 | |||
| AK032317.1 | BAC27810.1 | |||
| AK080619.1 | BAC37961.1 | |||
| BC108988.2 | AAI08989.1 | |||
| BC108989.2 | AAI08990.1 | |||
| FJ207475.1 | ||||
| FJ207476.1 |
More Information
Set Membership: |
|

Genomic Map