Assay Details
Target Gene Details
Entrez Gene ID: | 22065 |
Gene Name: | transient receptor potential cation channel, subfamily C, member 3 |
Gene Aliases: |
Mwk, Trcp3, Trp3, Trrp3 |
Location: |
Chr.3:36620482-36690229 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpc3 | NM_019510.2 | 3 | 1196 | NP_062383.2 |
| AF190645.1 | 2 | 786 | AAF01468.1 | |
| AK032317.1 | 3 | 1197 | BAC27810.1 | |
| AK080619.1 | 3 | 1184 | BAC37961.1 | |
| BC108988.2 | 2 | 785 | AAI08989.1 | |
| BC108989.2 | 2 | 786 | AAI08990.1 | |
| FJ207475.1 | 2 | 786 | ||
| FJ207476.1 | 2 | 786 |
More Information
Set Membership: |
|

Genomic Map