Assay Details
Target Gene Details
Entrez Gene ID: | 65973 |
Gene Name: | aspartate-beta-hydroxylase |
Gene Aliases: |
2310005F16Rik, 3110001L23Rik, AI848629, AW261690, AW561901, BAH, C79816, cI-37 |
Location: |
Chr.4:9449085-9669344 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asph | NM_001177849.1 | NP_001171320.1 | ||
| NM_001290367.1 | NP_001277296.1 | |||
| NM_023066.3 | NP_075553.2 | |||
| AF289486.1 | AAG40808.1 | |||
| AF289487.1 | AAG40809.1 | |||
| AK030293.1 | BAC26882.1 | |||
| AK035735.1 | BAC29171.1 | |||
| AK049506.1 | BAC33783.1 |
More Information
Set Membership: |
|

Genomic Map