Assay Details
Target Gene Details
Entrez Gene ID: | 22094 |
Gene Name: | thyroid stimulating hormone, beta subunit |
Gene Aliases: |
- |
Location: |
Chr.3:102773624-102803490 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tshb | NM_001165939.1 | NP_001159411.1 | ||
| NM_001165940.1 | NP_001159412.1 | |||
| NM_009432.2 | NP_033458.2 | |||
| BC116829.1 | ||||
| BC116831.1 | ||||
| BC144732.1 | ||||
| J00644.1 | ||||
| M35719.1 | AAA37307.1 | |||
| M35720.1 | AAA37308.1 | |||
| M35721.1 | AAA37309.1 | |||
| M54943.1 | AAA40492.1 |
More Information
Set Membership: |
|

Genomic Map