Assay Details
Target Gene Details
Entrez Gene ID: | 22094 |
Gene Name: | thyroid stimulating hormone, beta subunit |
Gene Aliases: |
- |
Location: |
Chr.3:102773624-102803490 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tshb | NM_001165939.1 | 3 | 158 | NP_001159411.1 |
| NM_001165940.1 | 2 | 111 | NP_001159412.1 | |
| NM_009432.2 | 4 | 199 | NP_033458.2 | |
| BC116829.1 | 3 | 120 | ||
| BC116831.1 | 3 | 120 | ||
| BC144732.1 | 2 | 78 | ||
| J00644.1 | 3 | 132 | ||
| M35719.1 | AAA37307.1 | |||
| M35720.1 | AAA37308.1 | |||
| M35721.1 | AAA37309.1 | |||
| M54943.1 | 3 | 132 | AAA40492.1 |
More Information
Set Membership: |
|

Genomic Map