Assay Details
Target Gene Details
Entrez Gene ID: | 71901 |
Gene Name: | family with sequence similarity 219, member A |
Gene Aliases: |
2310028H24Rik |
Location: |
Chr.4:41517691-41569533 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam219a | NM_001159583.1 | NP_001153055.1 | ||
| NM_027993.3 | NP_082269.2 | |||
| AK009522.1 | BAB26337.1 | |||
| AK134737.1 | ||||
| BC147808.1 | ||||
| BC147810.1 | ||||
| BC147834.1 | ||||
| BC147836.1 | ||||
| BC172167.1 | ||||
| BC172173.1 | ||||
| BY718591.1 |

Genomic Map