Assay Details
Target Gene Details
Entrez Gene ID: | 75691 |
Gene Name: | ankyrin repeat and sterile alpha motif domain containing 6 |
Gene Aliases: |
2210417J20Rik, Ankrd14, Gm635, SamCystin, Samd6, b2b1801.1Clo |
Location: |
Chr.4:47015365-47057335 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anks6 | NM_001024136.1 | NP_001019307.1 |
More Information
Set Membership: |
|

Genomic Map