Assay Details
Target Gene Details
Entrez Gene ID: | 65973 |
Gene Name: | aspartate-beta-hydroxylase |
Gene Aliases: |
2310005F16Rik, 3110001L23Rik, AI848629, AW261690, AW561901, BAH, C79816, cI-37 |
Location: |
Chr.4:9449085-9669344 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
More Information
Set Membership: |
|

Genomic Map