Assay Details
Target Gene Details
Entrez Gene ID: | 67628 |
Gene Name: | acidic (leucine-rich) nuclear phosphoprotein 32 family, member B |
Gene Aliases: |
2410015B15Rik, PAL31, PHAPI2a, Ssp29 |
Location: |
Chr.4:46451117-46472657 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anp32b | NM_130889.2 | NP_570959.1 | ||
| AB025582.1 | BAB12436.1 | |||
| AK011134.1 | ||||
| AK136648.1 | BAE23087.1 | |||
| AK165807.1 | BAE38387.1 | |||
| BC003489.1 | AAH03489.1 | |||
| BC005628.1 | AAH05628.1 | |||
| BC093506.1 | AAH93506.1 |
More Information
Set Membership: |
|

Genomic Map