Assay Details
Target Gene Details
Entrez Gene ID: | 69928 |
Gene Name: | apoptosis-inducing, TAF9-like domain 1 |
Gene Aliases: |
2610040C18Rik, 2810407L01Rik, CENP-S |
Location: |
Chr.4:149128349-149137600 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apitd1 | NM_027263.2 | NP_081539.1 | ||
| AK011725.1 | BAB27801.1 | |||
| AK012995.1 | BAC25385.1 | |||
| AK013038.1 | BAC25387.1 | |||
| BC048521.1 | AAH48521.1 | |||
| DV656255.1 |
More Information
Set Membership: |
|

Genomic Map