Assay Details
Target Gene Details
Entrez Gene ID: | 66077 |
Gene Name: | aurora kinase A interacting protein 1 |
Gene Aliases: |
0610033H09Rik, Aip, Akip, MRP-S38 |
Location: |
Chr.4:155831269-155833098 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aurkaip1 | NM_025338.4 | 1 | 383 | NP_079614.1 |
| AK002737.1 | BAB22317.1 | |||
| AK003442.1 | BAB22792.1 | |||
| AK033452.1 | BAC28297.1 | |||
| AK144719.1 | BAE26030.1 | |||
| AK166049.1 | BAE38543.1 | |||
| CX233396.1 | 1 | 383 |

Genomic Map