Assay Details
Target Gene Details
Entrez Gene ID: | 242594 |
Gene Name: | RIKEN cDNA 1700024P16 gene |
Gene Aliases: |
C1orf168, Gm620 |
Location: |
Chr.4:104833834-105016864 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700024P16Rik |
Target Gene Details
Entrez Gene ID: | 230558 |
Gene Name: | complement component 8, alpha polypeptide |
Gene Aliases: |
- |
Location: |
Chr.4:104815679-104876486 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8a | NM_001290645.1 | NP_001277574.1 | ||
| NM_146148.2 | NP_666260.1 | |||
| AB077295.1 | BAC41360.1 | |||
| AB077296.1 | BAC41361.1 | |||
| AB077297.1 | BAC41362.1 | |||
| AB077298.1 | BAC41363.1 | |||
| AK090245.1 | ||||
| BC027748.1 | AAH27748.1 |

Genomic Map