Assay Details
Target Gene Details
Entrez Gene ID: | 214444 |
Gene Name: | CDK5 regulatory subunit associated protein 2 |
Gene Aliases: |
2900018K03Rik, an, mKIAA1633 |
Location: |
Chr.4:70217690-70410461 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5rap2 | NM_145990.3 | NP_666102.2 | ||
| AK129411.1 | BAC98221.1 | |||
| BC059253.1 | AAH59253.1 | |||
| BC086696.1 |

Genomic Map