Assay Details
Target Gene Details
Entrez Gene ID: | 229096 |
Gene Name: | YTH domain family 3 |
Gene Aliases: |
9130022A11Rik |
Location: |
Chr.3:16183005-16217037 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ythdf3 | NM_001145919.1 | NP_001139391.1 | ||
| NM_172677.3 | NP_766265.3 | |||
| NR_027375.1 | ||||
| AK039176.1 | BAC30267.1 | |||
| AK053736.1 | BAC35498.1 | |||
| AK137245.1 | BAE23284.1 | |||
| BC057158.1 | AAH57158.1 | |||
| BC067040.1 | AAH67040.1 | |||
| BC067042.1 | AAH67042.1 | |||
| CJ066078.1 |
More Information
Set Membership: |
|

Genomic Map