Assay Details
Target Gene Details
Entrez Gene ID: | 74753 |
Gene Name: | RIKEN cDNA 5830415F09 gene |
Gene Aliases: |
AV014846, Nap1 |
Location: |
Chr.4:46376504-46393557 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5830415F09Rik | NM_029086.2 | 3 | 361 | NP_083362.1 |
| AK017931.1 | 3 | 372 | BAB31009.1 | |
| AK044073.1 | 3 | 355 | BAC31763.1 | |
| AK052584.1 | 3 | 376 | ||
| AK153576.1 | 3 | 362 | BAE32105.1 | |
| BC092543.1 | 3 | 362 | AAH92543.1 |

Genomic Map