Assay Details
Target Gene Details
Entrez Gene ID: | 74753 |
Gene Name: | RIKEN cDNA 5830415F09 gene |
Gene Aliases: |
AV014846, Nap1 |
Location: |
Chr.4:46376504-46393557 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5830415F09Rik | NM_029086.2 | NP_083362.1 | ||
| AK017931.1 | 5 | 1267 | BAB31009.1 | |
| AK044073.1 | 5 | 1250 | BAC31763.1 | |
| AK052584.1 | 5 | 1272 | ||
| AK153576.1 | BAE32105.1 | |||
| BC092543.1 | 5 | 1257 | AAH92543.1 |

Genomic Map