Assay Details
Target Gene Details
Entrez Gene ID: | 14595 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 1 |
Gene Aliases: |
B-1,4-GalT, B-1,4-GalT1, GalT, Ggtb, Ggtb2, b4Gal-T1, beta-1,4-GalT, beta-1,4-GalT1, beta4Gal-T1 |
Location: |
Chr.4:40804582-40854537 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4galt1 | NM_022305.4 | NP_071641.1 | ||
| AK029474.1 | ||||
| AK131721.1 | ||||
| AK132970.1 | BAE21450.1 | |||
| AK154393.1 | BAE32556.1 | |||
| AK167322.1 | BAE39424.1 | |||
| BC053006.1 | AAH53006.1 | |||
| D00314.1 | BAA00216.1 | |||
| J03880.1 | AAA37297.1 |
More Information
Set Membership: |
|

Genomic Map