Assay Details
Target Gene Details
Entrez Gene ID: | 214444 |
Gene Name: | CDK5 regulatory subunit associated protein 2 |
Gene Aliases: |
2900018K03Rik, an, mKIAA1633 |
Location: |
Chr.4:70217690-70410461 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5rap2 | NM_145990.3 | 27 | 4257 | NP_666102.2 |
| AK129411.1 | 27 | 4257 | BAC98221.1 | |
| BC059253.1 | 15 | 2848 | AAH59253.1 | |
| BC086696.1 | 27 | 4159 |

Genomic Map