Assay Details
Target Gene Details
Entrez Gene ID: | 99526 |
Gene Name: | ubiquitin specific peptidase 53 |
Gene Aliases: |
AA939927, Phxr3, Sp6, mKIAA1350 |
Location: |
Chr.3:122931486-122984509 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp53 | NM_133857.3 | 12 | 2080 | NP_598618.1 |
| AK036876.1 | 12 | 2081 | BAC29614.1 | |
| BC022221.1 | 2 | 222 | AAH22221.1 | |
| BC132339.1 | 11 | 1813 | ||
| BC145707.1 | 11 | 1813 |
More Information
Set Membership: |
|

Genomic Map