Assay Details
Target Gene Details
Entrez Gene ID: | 213234 |
Gene Name: | zinc finger, B-box domain containing |
Gene Aliases: |
4931432L23 |
Location: |
Chr.3:75037895-75165050 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zbbx | NM_172515.2 | NP_766103.2 | ||
AK029899.1 | ||||
AK133102.1 | BAE21512.1 | |||
BC050789.1 |
