Assay Details
Target Gene Details
Entrez Gene ID: | 99526 |
Gene Name: | ubiquitin specific peptidase 53 |
Gene Aliases: |
AA939927, Phxr3, Sp6, mKIAA1350 |
Location: |
Chr.3:122931486-122984509 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp53 | NM_133857.3 | 4 | 991 | NP_598618.1 |
| AK030317.1 | 5 | 1075 | BAC26897.1 | |
| AK036876.1 | 4 | 992 | BAC29614.1 | |
| AK040332.1 | 5 | 892 | ||
| AK045953.1 | 4 | 1054 | BAC32545.1 | |
| AK140453.1 | 5 | 954 | BAE24394.1 | |
| BC132339.1 | 3 | 724 | ||
| BC145707.1 | 3 | 724 |
More Information
Set Membership: |
|

Genomic Map