Assay Details
Target Gene Details
Entrez Gene ID: | 229055 |
Gene Name: | zinc finger and BTB domain containing 10 |
Gene Aliases: |
4832414A18, AU021107, RINZF |
Location: |
Chr.3:9250567-9285333 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb10 | NM_177660.3 | NP_808328.2 | ||
| AK048280.1 | ||||
| AK083443.1 | BAC38917.1 |
More Information
Set Membership: |
|

Genomic Map