Assay Details
Target Gene Details
Entrez Gene ID: | 545539 |
Gene Name: | predicted gene 15417 |
Gene Aliases: |
ENSMUSG00000074466, OTTMUSG00000021274 |
Location: |
Chr.3:89391864-89398779 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15417 | NR_040403.1 | |||
| AK161599.1 |
Target Gene Details
Entrez Gene ID: | 22724 |
Gene Name: | zinc finger and BTB domain containing 7B |
Gene Aliases: |
Thpok, Zfp67, c-Krox |
Location: |
Chr.3:89377644-89394814 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb7b | NM_009565.4 | NP_033591.2 | ||
| BC091651.1 | AAH91651.1 |
More Information
Set Membership: |
|

Genomic Map