Assay Details
Target Gene Details
Entrez Gene ID: | 67157 |
Gene Name: | RIKEN cDNA 2610301B20 gene |
Gene Aliases: |
AI428449 |
Location: |
Chr.4:10874249-10899425 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610301B20Rik | NM_026005.3 | NP_080281.3 | ||
| AK011950.1 | BAB27934.1 | |||
| AK146358.1 | BAE27110.1 | |||
| BC058777.1 | AAH58777.1 | |||
| BM948390.1 | ||||
| CX220600.1 |

Genomic Map