Assay Details
Target Gene Details
Entrez Gene ID: | 66451 |
Gene Name: | RIKEN cDNA 2610528J11 gene |
Gene Aliases: |
TEMP |
Location: |
Chr.4:118526995-118530217 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610528J11Rik | NM_025572.2 | NP_079848.1 | ||
| AK009128.1 | BAB26091.1 | |||
| AK012175.1 | BAB28079.1 | |||
| AK147115.1 | BAE27687.1 | |||
| BC022764.1 | AAH22764.1 |

Genomic Map