Assay Details
Target Gene Details
Entrez Gene ID: | 170753 |
Gene Name: | zinc finger protein 704 |
Gene Aliases: |
C030026M03, C030034L19Rik, Gig1, Znf704 |
Location: |
Chr.3:9427010-9610085 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp704 | NM_133218.2 | NP_573481.1 | ||
| AF292939.1 | AAG34027.1 | |||
| DT908776.1 |
More Information
Set Membership: |
|

Genomic Map