Assay Details
Target Gene Details
Entrez Gene ID: | 100206 |
Gene Name: | ADP-ribosylhydrolase like 2 |
Gene Aliases: |
AI836109, Arh3 |
Location: |
Chr.4:126316351-126321703 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adprhl2 | NM_133883.2 | 2 | 309 | NP_598644.1 |
| AJ427296.1 | 2 | 309 | CAD20317.1 | |
| AK143583.1 | 2 | 299 | BAE25451.1 | |
| AK147034.1 | 2 | 285 | BAE27626.1 | |
| AK149904.1 | 2 | 301 | ||
| AK150202.1 | 2 | 301 | ||
| AK169070.1 | 2 | 296 | BAE40857.1 | |
| AK171780.1 | 2 | 299 | BAE42660.1 | |
| BC023177.1 | 2 | 286 | ||
| BC045203.1 | 2 | 347 | AAH45203.1 |
More Information
Set Membership: |
|

Genomic Map