Assay Details
Target Gene Details
Entrez Gene ID: | 110198 |
Gene Name: | aldo-keto reductase family 7, member A5 (aflatoxin aldehyde reductase) |
Gene Aliases: |
0610025K21Rik, Afar, Afar1, Akr7a2 |
Location: |
Chr.4:139310738-139318786 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr7a5 | NM_025337.3 | NP_079613.3 | ||
| AF525358.2 | AAO38437.2 | |||
| AJ271800.1 | CAC81077.1 | |||
| AJ271801.1 | CAC81078.1 | |||
| AK002664.1 | ||||
| AK003915.1 | ||||
| AK143203.1 | BAE25302.1 | |||
| BC031857.1 | AAH31857.1 |
More Information
Set Membership: |
|

Genomic Map