Assay Details
Target Gene Details
Entrez Gene ID: | 242594 |
Gene Name: | RIKEN cDNA 1700024P16 gene |
Gene Aliases: |
C1orf168, Gm620 |
Location: |
Chr.4:104833834-105016864 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700024P16Rik | NM_001162980.1 | 5 | 1229 | NP_001156452.1 |
| NM_001285952.1 | 6 | 1307 | NP_001272881.1 | |
| NM_001285953.1 | 5 | 1162 | NP_001272882.1 | |
| NR_104376.1 | 5 | 1229 | ||
| AK028077.1 | 5 | 1169 | ||
| AK133762.1 | 5 | 1100 | ||
| BC137723.1 | 5 | 1229 | ||
| BC144857.1 | 6 | 1307 | ||
| BC144858.1 | 5 | 1230 |
More Information
Set Membership: |
|

Genomic Map