Assay Details
Target Gene Details
Entrez Gene ID: | 75986 |
Gene Name: | agmatine ureohydrolase (agmatinase) |
Gene Aliases: |
5033405N08Rik |
Location: |
Chr.4:141746675-141759263 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agmat | NM_001081408.1 | NP_001074877.1 | ||
| AK017155.1 | ||||
| BC025916.1 | AAH25916.1 | |||
| BC115693.2 |
More Information
Set Membership: |
|

Genomic Map